missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acebutolol hydrochloride, Thermo Scientific Chemicals
Cardioselective beta-adrenergic blocker
$133.32 - $467.51
Chemical Identifiers
CAS | 34381-68-5 |
---|---|
Molecular Formula | C18H29ClN2O4 |
Molecular Weight (g/mol) | 372.89 |
MDL Number | MFCD00078860 |
InChI Key | KTUFKADDDORSSI-UHFFFAOYNA-N |
Synonym | acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn |
PubChem CID | 441307 |
ChEBI | CHEBI:2380 |
IUPAC Name | hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride |
SMILES | [H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ6379003
|
Thermo Scientific Chemicals
J6379003 |
1 g |
Each for $133.32
|
|
||||||
AAJ6379006
|
Thermo Scientific Chemicals
J6379006 |
5 g |
Each for $215.49
|
|
||||||
AAJ6379009
|
Thermo Scientific Chemicals
J6379009 |
10 g |
Each for $467.51
|
|
||||||
Description
Cardioselective beta-adrenergic blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34381-68-5 | |
372.89 | |
KTUFKADDDORSSI-UHFFFAOYNA-N | |
441307 | |
hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride |
C18H29ClN2O4 | |
MFCD00078860 | |
acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
CHEBI:2380 | |
[H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O |
Specifications
34381-68-5 | |
White | |
MFCD00078860 | |
14,19 | |
KTUFKADDDORSSI-UHFFFAOYNA-N | |
hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride | |
441307 | |
372.89 | |
Acebutolol hydrochloride |
141°C to 143°C | |
C18H29ClN2O4 | |
1 g | |
acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn | |
[H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O | |
372.89 | |
CHEBI:2380 | |
Powder |
Safety and Handling
EINECSNumber : 251-980-3
RTECSNumber : ES5235000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only