missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetaldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 033244AY

 View more versions of this product

Catalog No. AA33244AY


Explore available promotions

Description

Description

Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetaldehyde
-128°C
0.780 g/mL
-27°C (-17°F)
C2H4O
MFCD00006991
UN1089
Air Sensitive
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
177
44.06
Liquid
75-07-0
Colorless
20.8°C
Pungent
1.3316
4 x 500 mL
505984
14,39
Miscible with water,ethanol,ether,benzene,toluene,xylene,turpentine,acetone and gasoline.Slightly miscible with chloroform.
CC=O
44.053
CHEBI:15343
98.5%

Chemical Identifiers

75-07-0
44.053
IKHGUXGNUITLKF-UHFFFAOYSA-N
177
acetaldehyde
C2H4O
MFCD00006991
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
CHEBI:15343
CC=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H351-H302-H319-H335
Extremely flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c

H224-H319-H335-H341-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ACETALDEHYDE

EINECSNumber : 200-836-8

RTECSNumber : AB1925000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only