Learn More
Acetaldehyde, 99%, Thermo Scientific Chemicals
$83.84 - $1025.66
Chemical Identifiers
CAS | 75-07-0 |
---|---|
Molecular Formula | C2H4O |
Molecular Weight (g/mol) | 44.053 |
MDL Number | MFCD00006991 |
InChI Key | IKHGUXGNUITLKF-UHFFFAOYSA-N |
Synonym | ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique |
PubChem CID | 177 |
ChEBI | CHEBI:15343 |
IUPAC Name | acetaldehyde |
SMILES | CC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA33244AE
|
Thermo Scientific Chemicals
033244AE |
100 mL |
Each for $83.84
|
|
|||||
AA33244AP
|
Thermo Scientific Chemicals
033244AP |
500 mL |
Each for $257.74
|
|
|||||
AA33244AY
|
Thermo Scientific Chemicals
033244AY |
4 x 500 mL |
Each for $1,025.66
|
|
|||||
Description
Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-07-0 | |
44.053 | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
177 | |
acetaldehyde |
C2H4O | |
MFCD00006991 | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
CHEBI:15343 | |
CC=O |
Specifications
75-07-0 | |
0.780 g/mL | |
-27°C (-17°F) | |
1.3316 | |
100 mL | |
505984 | |
14,39 | |
Miscible with water,ethanol,ether,benzene,toluene,xylene,turpentine,acetone and gasoline.Slightly miscible with chloroform. | |
CC=O | |
44.053 | |
CHEBI:15343 | |
98.5% | |
Acetaldehyde |
-128°C | |
20.8°C | |
C2H4O | |
MFCD00006991 | |
1089 | |
Air Sensitive | |
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique | |
IKHGUXGNUITLKF-UHFFFAOYSA-N | |
acetaldehyde | |
177 | |
44.06 | |
Liquid |
Safety and Handling
GHS H Statement
H224-H351-H302-H319-H335
Extremely flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
H224-H319-H335-H341-H350
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ACETALDEHYDE
EINECSNumber : 200-836-8
RTECSNumber : AB1925000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only