Learn More
Acetaldehyde ethyl propargyl acetal, 98%, Thermo Scientific™
$260.32 - $260.32
Chemical Identifiers
CAS | 18669-04-0 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00066344 |
InChI Key | QKBBTQJLUGADEG-UHFFFAOYSA-N |
PubChem CID | 86774 |
IUPAC Name | 3-(1-ethoxyethoxy)prop-1-yne |
SMILES | CCOC(C)OCC#C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC390640050
|
Thermo Scientific Chemicals
390640050 |
5 g | Glass bottle |
Each for $260.32
|
|
||||
Chemical Identifiers
18669-04-0 | |
128.17 | |
QKBBTQJLUGADEG-UHFFFAOYSA-N | |
3-(1-ethoxyethoxy)prop-1-yne |
C7H12O2 | |
MFCD00066344 | |
86774 | |
CCOC(C)OCC#C |
Specifications
18669-04-0 | |
100.0 | |
0.8930g/mL | |
Authentic | |
Glass bottle | |
1.4150 to 1.417 | |
5 g | |
3-1-ethoxyethoxy prop-1-yne, acetaldehyde ethyl propargyl acetal, 1-propyne, 3-1-ethoxyethoxy, 1-ethoxy-1-prop-2-yn-1-yloxy ethane, 3-1-ethoxyethoxy propyne, 1-ethoxyethoxy-2-propyne, 1-1-ethoxyethoxy-2propyne, 1-1-ethoxyethoxy-2-propyne, 5-methyl-4,6-dioxa-1-octyne, 1-propyne,3-1-ethoxyethoxy | |
CCOC(C)OCC#C | |
128.17 | |
128.17 | |
Liquid |
97.5 | |
Colorless to Yellow | |
141°C to 142°C | |
97.5% min. (GC) | |
C7H12O2 | |
MFCD00066344 | |
0.893 | |
QKBBTQJLUGADEG-UHFFFAOYSA-N | |
3-(1-ethoxyethoxy)prop-1-yne | |
86774 | |
98% | |
Acetaldehyde ethyl propargyl acetal |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 242-490-
RUO – Research Use Only