missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetanilide, 99+%, Thermo Scientific Chemicals
$63.59 - $965.40
Chemical Identifiers
CAS | 103-84-4 |
---|---|
Molecular Formula | C8H9NO |
Molecular Weight (g/mol) | 135.17 |
MDL Number | MFCD00008674 |
InChI Key | FZERHIULMFGESH-UHFFFAOYSA-N |
Synonym | acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene |
PubChem CID | 904 |
ChEBI | CHEBI:28884 |
IUPAC Name | N-phenylacetamide |
SMILES | CC(=O)NC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC150810050
|
Thermo Scientific Chemicals
150810050 |
5 g | Glass bottle |
Each for $63.59
|
|
|||||
AC150812500
|
Thermo Scientific Chemicals
150812500 |
250 g | Plastic bottle |
Each for $99.00
|
|
|||||
AC150815000
|
Thermo Scientific Chemicals
150815000 |
500 g | Plastic bottle |
Each for $131.19
|
|
|||||
AC150810010
|
Thermo Scientific Chemicals
150810010 |
1 kg | Plastic bottle |
Each for $213.30
|
|
|||||
AC150810051
|
Thermo Scientific Chemicals
150810051 |
5 kg | Plastic drum |
Each for $965.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-84-4 | |
135.17 | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
904 | |
N-phenylacetamide |
C8H9NO | |
MFCD00008674 | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
CHEBI:28884 | |
CC(=O)NC1=CC=CC=C1 |
Specifications
103-84-4 | |
100.0 | |
Blue-Gray to White | |
304.0°C | |
≥111°C to ≤115°C | |
99% min. (GC) | |
C8H9NO | |
MFCD00008674 | |
12, 237 | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
N-phenylacetamide | |
904 | |
135.17 | |
99+% | |
Acetanilide |
99.0 | |
111.0°C to 115.0°C | |
4.65 (Air = 1.0) | |
173°C | |
Conforms to Reference | |
Glass bottle | |
CH3CONHC6H5 | |
5 g | |
15, 49 | |
Solubility in water: slightly soluble (5g/L at 25°C). Other solubilities: 1g/20mL boiling water,1g/3.4mL alcohol,1g/4mL ac,etone,1g/3.7mL chloroform,1g/5mL glycerol,1g/8mL,dioxane,1g/18mL diethyl ether,1g/47mL benzene,,sparingly soluble in pe | |
CC(=O)NC1=CC=CC=C1 | |
135.17 | |
CHEBI:28884 | |
13hPa at 114°C | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 203-150-7
RUO – Research Use Only