Learn More
Acetic Acid, Glacial (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific A38P20
Specifications
Acetic Acid, Glacial | |
16°C | |
117°C | |
Assay: ≥99.7% w/w | |
CH3COOH | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Certified ACS | |
20 L | |
Poly Drum | |
8ppm max. |
2.5 | |
64-19-7 | |
Colorless | |
C2H4O2 | |
MFCD00036152 | |
15, 54 | |
Soluble in water | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% (w/w) | |
1.048g/cm³ | |
40°C | |
Liquid | |
1.52 kPa at 20°C |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 8: Corrosive
Recommended Storage : RT