Learn More
Acetic Acid, Glacial (Certified ACS), Fisher Chemical™
$209.46 - $18046.08
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A38-500
|
Thermo Fisher Scientific
FLA38500 |
500 mL | Glass Bottle |
Each for $209.46
Case of 6 Each for $1,256.76
|
|
||||
A38S-500
|
Thermo Fisher Scientific
FLA38S500 |
500 mL | Safe-Cote™ Glass |
Each for $294.85
Case of 6 Each for $1,769.10
|
|
||||
A38-212
|
Thermo Fisher Scientific
FLA38212 |
2.5 L | Glass Bottle |
Each for $406.31
|
|
||||
A38P-20
|
Thermo Fisher Scientific
A38P20 |
20 L | Poly Drum |
Each for $2,202.23
|
|
||||
A38-450LB
|
Thermo Fisher Scientific
FLA38450LB |
450 lb. | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
2.5 | |
64-19-7 | |
Colorless | |
C2H4O2 | |
MFCD00036152 | |
15, 54 | |
Soluble in water | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% (w/w) | |
1.048g/cm³ | |
40°C | |
Liquid | |
1.52 kPa at 20°C |
16°C | |
117°C | |
Assay: ≥99.7% w/w | |
CH3COOH | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Certified ACS | |
500 mL | |
Glass Bottle | |
8ppm max. | |
Acetic Acid, Glacial |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 8: Corrosive
Recommended Storage : RT