missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid, Glacial (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA38S500

 View more versions of this product

Catalog No. A38S-500



Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid, Glacial
16°C
117°C
Assay: ≥99.7% w/w
CH3COOH
2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Certified ACS
500 mL
Safe-Cote™ Glass
8ppm max.
2.5
64-19-7
Colorless
C2H4O2
MFCD00036152
15, 54
Soluble in water
CC(O)=O
60.05
CHEBI:15366
≥99.7% (w/w)
1.048g/cm³
40°C
Liquid
1.52 kPa at 20°C

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 8: Corrosive

Recommended Storage : RT

SDS Documents

Documents

Accessories
Promotions

Promotions