missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Acid, Glacial (Certified ACS Plus), Fisher BioReagents
Supplier: Fisher BioReagents BP2401C212
Description
Acetic Acid, Glacial can be used for several biochemical procedures. Some products are packed in Safe Cote™ which give additional safety for chemical and user both.Specifications
Acetic Acid, Glacial | |
16°C | |
117°C | |
C2H4O2 | |
MFCD00036152 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
Certified ACS Plus | |
6 x 2.5 L | |
Liquid | |
1.52 kPa at 20°C |
2.5 | |
64-19-7 | |
Colorless | |
CH3COOH | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% | |
1.048g/cm³ | |
Glass | |
8ppm max. |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive
Recommended Storage : RT