missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid, Glacial (Certified ACS Plus), Fisher BioReagents

$193.50 - $787.18

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP2401-500
SDS View Documents Promotion Details
Fisher BioReagents
BP2401500
500 mL Glass
Each for $254.68
Case of 6 Each for $1,528.08
 
BP2401-212
SDS Accessories View Documents
Fisher BioReagents
BP2401212
2.5 L Glass
Each for $193.50
 
BP2401C-212
SDS View Documents Promotion Details
Fisher BioReagents
BP2401C212
6 x 2.5 L Glass
Case of 6 Each for $787.18
 
Description

Description

Acetic Acid, Glacial can be used for several biochemical procedures. Some products are packed in Safe Cote™ which give additional safety for chemical and user both.
Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

2.5
64-19-7
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
≥99.7%
1.048g/cm³
Glass
8ppm max.
Acetic Acid, Glacial
16°C
117°C
C2H4O2
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Certified ACS Plus
500 mL
Liquid
1.52 kPa at 20°C
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

Recommended Storage : RT

SDS
missing translation for 'documents'

missing translation for 'documents'