missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing


Acetic Acid, Glacial (HPLC), Fisher Chemical™
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
Supplier: Fisher Chemical FLA35500
Description
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
Specifications
0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1.0 AU max. at 255nm | |
2.5 | |
16°C | |
117°C | |
Color (APHA) 10 | |
≥99.7 w/w % CH3COOH | |
CH3COOH | |
2789 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05 | |
HPLC | |
500 mL | |
Liquid | |
1.52 kPa at 20°C |
Acetic Acid, Glacial | |
17.4N | |
64-19-7 | |
Colorless | |
HPLC | |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
≥99.7% (w/w) | |
1.048g/cm³ | |
Glass Bottle | |
0.001% |
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Safety and Handling


DOTInformation : Class 8: Corrosive