missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetoacetamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 442541000

 View more versions of this product

Catalog No. AC442541000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetoacetamide
96.0
-53°C to 56°C
113°C
96% min. (HPLC)
C4H7NO2
100 g
Solubility in water: miscible
CC(=O)CC(=O)N
101.11
CHEBI:28515
97%
5977-14-0
100.0
White to Yellow
Authentic
Glass bottle
CH3C(O)CH2C(O)NH2
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
GCPWJFKTWGFEHH-UHFFFAOYSA-N
3-oxobutanamide
80077
101.11
Crystalline Powder

Chemical Identifiers

5977-14-0
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
CHEBI:28515
CC(=O)CC(=O)N
GCPWJFKTWGFEHH-UHFFFAOYSA-N
80077
3-oxobutanamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-774-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only