Learn More
Acetone, 99.6%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423240250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetone | |
67-64-1 | |
100.0 | |
0.7900g/mL | |
25 L | |
Metal Drum | |
1.3585 | |
99.6% | |
0.0003 meq/g max. | |
0.0006 meq/g max. | |
MFCD00008765 | |
0.32 mPa.s (20°C) | |
0.5% max. (K.F.) | |
02,13; 03,4; 06,9 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
ACS Reagent |
-95.0°C | |
99.5 | |
>4°C | |
56.0°C | |
−20°C | |
Liquid | |
0.001% max. | |
0.79 | |
C3H6O | |
CH3COCH3 | |
Vapor Pressure: 247mbar at 20°C | |
1090; Colorless | |
01, 635 | |
15, 65 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
99.6% |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only