missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, 99.6%, ACS reagent, Thermo Scientific Chemicals

$95.50 - $1342.92

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423245000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
423245000
500 mL EcoSafPak™ Glass Bottle
Each for $95.50
Case of 6 Each for $0.00
 
AC423240025
SDS View Documents
Thermo Scientific Chemicals
423240025
2.5 L Glass Bottle
Each for $219.70
 
AC423240040
SDS View Documents
Thermo Scientific Chemicals
423240040
4 L EcoSafPak™ Glass Bottle
Each for $347.21
Case of 4 Each for $0.00
 
AC423240250
SDS View Documents
Thermo Scientific Chemicals
423240250
25 L Metal Drum
Each for $1,342.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

-95.0°C
99.5
>4°C
56.0°C
-20°C (-4°C)
Liquid
0.001% max.
0.79
C3H6O
CH3COCH3
Vapor Pressure: 247mbar at 20°C
1090; Colorless
01, 635
15, 65
Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
CC(C)=O
58.08
CHEBI:15347
99.6%
Acetone
67-64-1
100.0
0.7900g/mL
500 mL
EcoSafPak™ Glass Bottle
1.3585
99.6%
0.0003 meq/g max.
0.0006 meq/g max.
MFCD00008765
0.32 mPa.s (20°C)
0.5% max. (K.F.)
02,13; 03,4; 06,9
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only