missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, ACS Reagent Grade, Ricca Chemical

$292.03 - $521.25

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RSOA00104A
View Documents
Ricca Chemical Company
RSOA00104A
4 L Natural Poly Bottle
Each for $292.03
 
RSOA001020F
View Documents
Ricca Chemical Company
RSOA001020F
20 L Cubitainer
Each for $521.25
 
Description

Description

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
CHEBI:15347
CC(C)=O
C3H6O
MFCD00008765
180
propan-2-one

Specifications

67-64-1
100.0
Natural Poly Bottle
max. 0.0003 meq/g
max. 0.0006 meq/g
max. 0.5%
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
min. 99.5%
98.0
4 L
max. 0.001%
C3H6O
MFCD00008765
acetone
CC(C)=O
58.08
CHEBI:15347
ACS
Videos
Safety and Handling

Safety and Handling

ShelfLife : 60 months

SDS
missing translation for 'documents'

missing translation for 'documents'