missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RSOA001020F
Specifications
| Acetone | |
| 98.0 | |
| 56°C | |
| Cubitainer | |
| max. 0.0003 meq/g | |
| max. 0.0006 meq/g | |
| max. 0.5% | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 99.5% |
| 67-64-1 | |
| 100.0 | |
| 20 L | |
| max. 0.001% | |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| ACS |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| CHEBI:15347 | |
| CC(C)=O |
| C3H6O | |
| MFCD00008765 | |
| 180 | |
| propan-2-one |
Safety and Handling
ShelfLife : 60 months