missing translation for 'onlineSavingsMsg'
Learn More

Acetone (Certified ACS), Fisher Chemical™

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

$251.36 - $12305.36

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A18-1
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA181
1 L HDPE Bottle
Each for $251.36
Case of 6 Each for $1,508.16
Only null left
 
A18-4
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA184
4 L Glass Bottle
Each for $422.82
Case of 4 Each for $1,691.28
Only null left
 
A18P-4
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA18P4
4 L Poly Bottle
Each for $341.36
Case of 4 Each for $1,365.44
Only null left
 
A18-20
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA1820
20 L Steel Pail
Each for $1,422.91
Only null left
 
A18-200
SDS Accessories View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA18200
200 L Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

7
67-64-1
>4°C
56°C
-20°C
Conforms to reference
Liquid
Assay: ≥99.5%
0.0003mEq/g max.
0.0006mEq/g max.
247mbar at 20°C
1090
15, 65
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
Certified ACS Reagent
-95°C
Colorless
0.790g/cm³
1 L
Pass Test
HDPE Bottle
0.001% max.
0.79
C3H6O
MFCD00008765
0.32 mPa/s at 20°C
0.5% max.
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
≥99.5%
Acetone
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'