Learn More
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
$524.25 - $7309.75
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A996-1
|
Thermo Fisher Scientific
FLA9961 |
1 L | Wide Neck Amber Bottle |
Each for $524.25
Case of 6 Each for $3,145.50
|
|
||||
A996-4
|
Thermo Fisher Scientific
BPA9964 |
4 L | Amber Soda Lime Glass |
Each for $1,090.50
Case of 4 Each for $4,362.00
|
|
||||
A996SK4
|
Thermo Fisher Scientific
A996SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $1,122.50
Case of 4 Each for $4,490.00
|
|
||||
A996N219
|
Thermo Fisher Scientific
A996N219 |
19 L | FisherPak |
Each for $7,309.75
|
|
||||
Description
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
- Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190-254nm range, to reduce interferences for smoother baselines
- Tested for HPLC gradient suitability
- Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
- Also meets ACS specifications
- Intended for research use only
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
-46°C | |
0.005 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm | |
>500°C | |
81°C | |
49°C (120.2°F) | |
Wide Neck Amber Bottle | |
1.3405 to 1.3425 (at 25°C) | |
Assay: 99.9% min by GC | |
C2H3N | |
MFCD00001878 | |
0.36 cP at 20°C | |
0.01% max. | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
High purity mobile phase for HPLC and/or extraction solvent for GC applications |
75-05-8 | |
Colorless | |
0.781g/cm³ | |
1 L | |
Pass Test | |
Liquid | |
0.8ppm max. | |
0.008mEq/g max. | |
0.0006mEq/g max. | |
97mbar at 20°C | |
1648; Colorless | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.9% | |
Acetonitrile |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid