missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl Sulfide 95.0+%, TCI America™

Supplier:  TCI America A09765G

 View more versions of this product

Catalog No. A09765G


Explore available promotions

Description

Description

Specifications

Specifications

Acetyl Sulfide
Yellow
C4H6O2S
5 g
Diacetyl Sulfide
CC(=O)SC(=O)C
118.15
118.15
Liquid
3232-39-1
120°C
MFCD00008753
1993
DPCQJLQPDJPRCM-UHFFFAOYSA-N
S-acetyl ethanethioate
76708
≥95.0% (GC)

Chemical Identifiers

3232-39-1
118.15
DPCQJLQPDJPRCM-UHFFFAOYSA-N
76708
CC(=O)SC(=O)C
C4H6O2S
MFCD00008753
Diacetyl Sulfide
S-acetyl ethanethioate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions