missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl Sulfide 95.0+%, TCI America™
$416.73 - $1384.48
Chemical Identifiers
CAS | 3232-39-1 |
---|---|
Molecular Formula | C4H6O2S |
Molecular Weight (g/mol) | 118.15 |
MDL Number | MFCD00008753 |
InChI Key | DPCQJLQPDJPRCM-UHFFFAOYSA-N |
Synonym | Diacetyl Sulfide |
PubChem CID | 76708 |
IUPAC Name | S-acetyl ethanethioate |
SMILES | CC(=O)SC(=O)C |
Chemical Identifiers
3232-39-1 | |
118.15 | |
DPCQJLQPDJPRCM-UHFFFAOYSA-N | |
76708 | |
CC(=O)SC(=O)C |
C4H6O2S | |
MFCD00008753 | |
Diacetyl Sulfide | |
S-acetyl ethanethioate |
Specifications
3232-39-1 | |
120°C | |
MFCD00008753 | |
1993 | |
DPCQJLQPDJPRCM-UHFFFAOYSA-N | |
S-acetyl ethanethioate | |
76708 | |
≥95.0% (GC) | |
Acetyl Sulfide |
Yellow | |
C4H6O2S | |
5 g | |
Diacetyl Sulfide | |
CC(=O)SC(=O)C | |
118.15 | |
118.15 | |
Liquid |
Safety and Handling
TSCA : No