missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acid Black 1 97.0+%, TCI America™

$73.08 - $657.68

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, parent, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A058625G
View Documents
TCI America
A058625G
25 g N/A
 
A0586100G
View Documents
TCI America
A0586100G
100 g N/A
 
A0586500G
View Documents
TCI America
A0586500G
500 g N/A
 
Description

Description

Specifications

Chemical Identifiers

1064-48-8
616.487
HKBVRFLHNUEVRO-UHFFFAOYSA-L
44134531
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C22H14N6Na2O9S2
MFCD00004017
amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, parent, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate

Specifications

1064-48-8
MFCD00004017
amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, parent, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
616.487
616.49
Crystalline Powder
C22H14N6Na2O9S2
25 g
HKBVRFLHNUEVRO-UHFFFAOYSA-L
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44134531
≥97.0% (HPLC)
Acid Black 1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1875&(5)-5232

RTECSNumber : QJ6196000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'