missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acridine Orange, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1315914

 View more versions of this product

Catalog No. AAL1315914


Explore available promotions

Description

Description

Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Acridine Orange
C17H20Cl3N3Zn
25 g
Hygroscopic
Soluble in water.
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
438.10
438.1
10127-02-3
MFCD00081043,MFCD00081043,MFCD00081043
3734978
acridine orange c.i. 46005
VADJQOXWNSPOQA-UHFFFAOYSA-L
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
71774334

Chemical Identifiers

10127-02-3
438.10
VADJQOXWNSPOQA-UHFFFAOYSA-L
71774334
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
C17H20Cl3N3Zn
MFCD00081043,MFCD00081043,MFCD00081043
acridine orange c.i. 46005
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H341

EINECSNumber : 233-353-6

RTECSNumber : AR7600000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only