missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acridine Orange, Thermo Scientific Chemicals

$58.99 - $132.22

Chemical Identifiers

CAS 10127-02-3
Molecular Formula C17H20Cl3N3Zn
Molecular Weight (g/mol) 438.10
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Synonym acridine orange c.i. 46005
PubChem CID 71774334
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1315906
View Documents
Thermo Scientific Chemicals
L1315906
5 g
Each for $58.99
 
AAL1315914
View Documents
Thermo Scientific Chemicals
L1315914
25 g
Each for $132.22
 
Description

Description

Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

10127-02-3
438.10
VADJQOXWNSPOQA-UHFFFAOYSA-L
71774334
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
C17H20Cl3N3Zn
MFCD00081043,MFCD00081043,MFCD00081043
acridine orange c.i. 46005
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride

Specifications

10127-02-3
MFCD00081043,MFCD00081043,MFCD00081043
3734978
acridine orange c.i. 46005
VADJQOXWNSPOQA-UHFFFAOYSA-L
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
71774334
Acridine Orange
C17H20Cl3N3Zn
5 g
Hygroscopic
Soluble in water.
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
438.10
438.1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H341

EINECSNumber : 233-353-6

RTECSNumber : AR7600000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only