Learn More
Acriflavine neutral, 13.3-15.8% chlorine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 211400250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acriflavine neutral | |
179°C to 181°C | |
8% max. (105°C, 2 hrs) | |
Glass bottle | |
MFCD00064307 | |
22, I, 650 | |
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine | |
PEJLNXHANOHNSU-UHFFFAOYSA-N | |
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride | |
469.03 | |
Powder |
8048-52-0 | |
Brown-Red to Orange | |
Authentic | |
C27H25ClN6 | |
25 g | |
15, 115 | |
Solubility in water: 1g/3mL. Other solubilities: incompletely soluble in alcohol,practically insoluble in ether,chloroform and,fixed oils | |
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 | |
468.99 | |
13.3 to 15.8% (chlorine) (on dried substance) |
Chemical Identifiers
8048-52-0 | |
468.99 | |
PEJLNXHANOHNSU-UHFFFAOYSA-N | |
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride |
C27H25ClN6 | |
MFCD00064307 | |
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine | |
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Warning
RTECSNumber : AR9660000
RUO – Research Use Only