missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine neutral, 13.3-15.8% chlorine

CAS: 8048-52-0 | C27H25ClN6 | 468.99 g/mol

$133.07 - $449.09

Chemical Identifiers

CAS 8048-52-0
Molecular Formula C27H25ClN6
Molecular Weight (g/mol) 468.99
MDL Number MFCD00064307
InChI Key PEJLNXHANOHNSU-UHFFFAOYSA-N
Synonym Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine
IUPAC Name 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
SMILES [Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211400250
SDS View Documents
Thermo Scientific Chemicals
211400250
25 g Glass bottle
Each for $133.07
Only null left
 
AC211401000
SDS View Documents
Thermo Scientific Chemicals
211401000
100 g Glass bottle
Each for $449.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8048-52-0
468.99
PEJLNXHANOHNSU-UHFFFAOYSA-N
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
C27H25ClN6
MFCD00064307
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Specifications

8048-52-0
Brown-Red to Orange
Authentic
C27H25ClN6
25 g
15, 115
Solubility in water: 1g/3mL. Other solubilities: incompletely soluble in alcohol,practically insoluble in ether,chloroform and,fixed oils
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
468.99
13.3 to 15.8% (chlorine) (on dried substance)
Acriflavine neutral
179°C to 181°C
8% max. (105°C, 2 hrs)
Glass bottle
MFCD00064307
22, I, 650
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine
PEJLNXHANOHNSU-UHFFFAOYSA-N
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
469.03
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Warning

RTECSNumber : AR9660000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only