Superior Glove™ Action™ Aramid Fiber String Knit Cut-Resistant Gloves
Aramid fiber cut and slash-resistant gloves feature leather palm and thumb to offer protection and extended wear.
Supplier: Superior Glove™ SKLPXL
Description
- 7-gauge
- Leather palms wrap around sides of fingers
- Leather thumb
- Durable tri-tan leather offers greater protection, longer wear
- Shell made of aramid fiber for cut and slash resistance
- Flame-resistance qualities
- Palms attached to knit shell using sewing thread made with Kevlar™ aramid fiber for added seam integrity
Specifications
Yellow, Gray | |
Aramid Fiber, Split Leather | |
X-Large | |
Knit |
ANSI A2 | |
Cut-resistant Gloves | |
sheet Metal, Metal stamping, Glass handling | |
SKLP/XL |