missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Superior Glove™ Action™ Aramid Fiber String Knit Cut-Resistant Gloves

Aramid fiber cut and slash-resistant gloves feature leather palm and thumb to offer protection and extended wear.

$28.99 - $34.49

Specifications

Color Yellow, Gray
Cut Resistance Rating ANSI A2
Material Aramid Fiber, Split Leather
Product Type Cut-resistant Gloves
For Use With (Application) sheet Metal, Metal stamping, Glass handling
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-635-5208
View Documents
Superior Glove™
SKLPS
Small
Pair for $28.99
Case of 12 Pair for $316.20
 
29-635-5209
View Documents
Superior Glove™
SKLPM
Medium
Pair for $31.44
Case of 12 Pair for $342.96
 
29-635-3220
View Documents
Superior Glove™
SKLPL
Large
Pair for $31.44
Case of 12 Pair for $342.96
 
29-635-5210
View Documents
Superior Glove™
SKLPXL
X-Large
Pair for $34.49
Case of 12 Pair for $376.20
 
Description

Description

  • 7-gauge
  • Leather palms wrap around sides of fingers
  • Leather thumb
  • Durable tri-tan leather offers greater protection, longer wear
  • Shell made of aramid fiber for cut and slash resistance
  • Flame-resistance qualities
  • Palms attached to knit shell using sewing thread made with Kevlar™ aramid fiber for added seam integrity
Specifications

Specifications

Yellow, Gray
Aramid Fiber, Split Leather
sheet Metal, Metal stamping, Glass handling
ANSI A2
Cut-resistant Gloves
Knit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'