Synthware™ Enlarging Adapters
With standard taper joints at top and bottom
Supplier:  Synthware™ A252419
 View more versions of this product
Catalog No. 31500515
Explore available promotions
Description
Description
Synthware enlarging adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is smaller in size than the top outer joint.
Specifications
Specifications
Organic Synthesis
Enlarging Adapter
Top joint: 24/40; Bottom joint: 19/22
Borosilicate Glass (3.3)
1