Synthware™ Enlarging Adapters
With standard taper joints at top and bottom
Supplier: Synthware™ A252924
Description
Synthware enlarging adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is smaller in size than the top outer joint.Specifications
Organic Synthesis | |
Enlarging Adapter | |
Top joint: 29/42; Bottom joint: 24/40 |
Borosilicate Glass (3.3) | |
1 |