Learn More
Thermo Scientific Chemicals Adenine, 99%
$143.96 - $1453.84
Chemical Identifiers
CAS | 73-24-5 |
---|---|
Molecular Formula | C5H5N5 |
Molecular Weight (g/mol) | 135.13 |
MDL Number | MFCD00041790 |
InChI Key | GFFGJBXGBJISGV-UHFFFAOYSA-N |
Synonym | adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine |
PubChem CID | 190 |
ChEBI | CHEBI:16708 |
IUPAC Name | 7H-purin-6-amine |
SMILES | NC1=C2NC=NC2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1490614
|
Thermo Scientific Chemicals
A1490614 |
25 g |
Each for $143.96
|
|
|||||
AAA1490618
|
Thermo Scientific Chemicals
A1490618 |
50 g |
Each for $252.21
|
|
|||||
AAA1490630
|
Thermo Scientific Chemicals
A1490630 |
250 g |
Each for $898.29
|
|
|||||
AAA1490636
|
Thermo Scientific Chemicals
A1490636 |
500 g |
Each for $1,453.84
|
|
|||||
Description
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.
Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
73-24-5 | |
135.13 | |
GFFGJBXGBJISGV-UHFFFAOYSA-N | |
190 | |
7H-purin-6-amine |
C5H5N5 | |
MFCD00041790 | |
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
CHEBI:16708 | |
NC1=C2NC=NC2=NC=N1 |
Specifications
73-24-5 | |
C5H5N5 | |
25 g | |
5777 | |
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
NC1=C2NC=NC2=NC=N1 | |
135.13 | |
CHEBI:16708 | |
99% |
99% | |
MFCD00041790 | |
UN2811 | |
14,152 | |
GFFGJBXGBJISGV-UHFFFAOYSA-N | |
7H-purin-6-amine | |
190 | |
135.13 | |
Adenine |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-796-1
RTECSNumber : AU6125000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only