missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adenosine, 99.8%, MP Biomedicals™

A nucleotide resulting from the union of one molecule of sugar -- d-ribose -- with a base, l-adenine

Supplier:  MP Biomedicals Inc 0210019905

 View more versions of this product

Catalog No. ICN10019905


Explore available promotions

Description

Description

Specifications

Specifications

Adenosine
White
99.80%
MFCD00005752
adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
267.25
CHEBI:16335
Crystalline Powder
58-61-7
2.09g/cm3
C10H13N5O4
5 g
OIRDTQYFTABQOQ-KQYNXXCUSA-N
(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
60961
267.2

Chemical Identifiers

58-61-7
267.25
OIRDTQYFTABQOQ-KQYNXXCUSA-N
60961
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
C10H13N5O4
MFCD00005752
adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
CHEBI:16335
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-389-9

SDS
Documents

Documents

Promotions

Promotions