Fisherbrand™ Adjustable Height Heavy Duty Steel Table with Locking Swivel Casters, Epoxy Work Surface
Fisherbrand™ Heavy Duty Steel Tables are great for general lab use.
 View more versions of this product
Catalog No. 06000825
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Comes with 4 locking swivel casters.
  • Steel frame is 2 x 2 in. tubular steel frame construction
  • Adjustable Height Heavy Duty Tables with casters are rated for 960 lbs UDL
Specifications
Specifications
Table Frame, Epoxy Work Surface and Locking Swivel Casters
2 years
76.2 to 91.4 cm
Fully Welded 2 in. x 2 in. Tubular Steel
152.4 cm
960 lb.
60 in.
36 in.
91.4 cm
435.4 kg
30 to 36 in.
White