Advanced Instruments Osmometer Standards
For in vitro diagnostic use
Supplier:  Advanced Instruments 3MA020
 View more versions of this product
Catalog No. 22046608
Explore available promotions
Description
Description
For in vitro diagnostic use
  • 2-Hydroxybiphenyl sodium salt tetrahydrate
  • 1% added as preservative
Specifications
Specifications
2°C to 30°C
3250, 3900 and A2O Osmometers
2- Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01% added as preservative
10 x 2 mL
200 mOsm/kg H2O
Osmometer Calibration Standard
To check the performance of your osmometer and test methods, in-vitro diagnostic use