METTLER TOLEDO Advanced MX Analytical Balance
METTLER TOLEDO Advanced MX Analytical Balances offer a brand new feature, the Backlit Draft Shield (5-place models only). Brightness extends over the draft shield's entire back panel providing exceptional visibility of the work area for highly precise dosing.
Supplier:  METTLER TOLEDO 30665091
 View more versions of this product
Catalog No. 01-804-330
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The stable SmartPan and high resolution weigh cell and FACT automatic internal adjustment ensure consistently accurate results. Efficient operation within the large, 7-inch touchscreen includes 8 built-in applications and statistical data analysis. When you're not using the balance, enable the power-saving mode to reduce energy usage and save costs.
All MX Analytical Balances include:
  • Backlit Draft Shield (5-place models only)
  • FACT automatic internal adjustment
  • 7-inch touchscreen
  • 8 built-in applications
  • Statistical data analysis
  • ErgoDoor
  • User management
  • Leveling assistant
  • StatusLight
  • 3 interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The MX Balance is the successor to the MS Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications
Specifications
USB-A, USB-C, Ethernet
Color Touchscreen
Metal
2 sec.
100/240 V, 50/60 Hz
42 g, 120 g
0.01 mg, 0.1 mg
MX105DU
Analytical Balance
No