Air-Tite™ All-Plastic Henke-Ject™ Syringes
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Henke-Ject™ syringes are available compatible with a variety of needle styles. Needles are sold separately.
Supplier: Air-Tite™ AL3HJ
Description
These Air-Tite, all plastic syringes are available compatible with a variety of needle styles. (Needles are sold separately.) Ideal for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures.
- Needle tips not included and must be purchased separately
- Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
- Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
- Luer Tip, Luer-Slip, or catheter tip compatibility
- Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
- Positive safety stop prevents accidental spills
- FDA registered
Order Info
Individually wrapped in convenient sterile strip packaging.
Specifications
FDA, CE, ISO 9001, DEHP Free | |
For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures | |
Henke-Ject™ Syringe | |
3 mL | |
Cone | |
Yes |
Plastic | |
Yes | |
0.1 cc | |
Norm-Ject | |
Luer Lock | |
Sterile |