Air-Tite™ All-Plastic Henke-Ject™ Syringes
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Henke-Ject™ syringes are available compatible with a variety of needle styles. Needles are sold separately.
$41.97 - $1571.59
Specifications
Certifications/Compliance | FDA, CE, ISO 9001, DEHP Free |
---|---|
Material | Plastic |
For Use With (Application) | For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures |
Latex Free | Yes |
Product Type | Henke-Ject™ Syringe |
Catalog Number | Mfr. No. | Volume (Metric) | Graduations | Connection Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Graduations | Connection Type | Price | Quantity | |||
14-817-43
|
Air-Tite™
AB10LS |
10 mL | 0.05 cc | Eccentric Catheter |
Case of 2000 Each for $1,571.59
|
|
|||
14-817-27
|
Air-Tite™
AL3HJ |
3 mL | 0.1 cc | Luer Lock |
Pack of 100 Each for $82.66
Case of 25 Pack for $2,066.50
|
|
|||
14-817-29
|
Air-Tite™
AL5HJ |
5 mL | 0.2 cc | Luer Tip |
Pack of 100 Each for $55.08
Case of 20 Pack for $1,101.60
|
|
|||
14-817-33
|
Air-Tite™
AL20HJ |
20 mL | 1 cc | Luer Tip |
Pack of 100 Each for $169.31
Case of 8 Pack for $1,354.48
|
|
|||
14-817-25
|
Air-Tite™
A1HJ |
1 mL | 0.05 cc | Leur Slip |
Pack of 100 Each for $49.32
Case of 18 Pack for $887.76
|
|
|||
14-817-26
|
Air-Tite™
A3HJ |
3 mL | 0.1 cc | Centric Luer |
Pack of 100 Each for $41.97
Case of 25 Pack for $1,049.25
|
|
|||
14-817-31
|
Air-Tite™
AL10HJ |
10 mL | 0.5 cc | Luer Tip |
Pack of 100 Each for $60.66
Case of 12 Pack for $727.92
|
|
|||
14-817-28
|
Air-Tite™
A5HJ |
5 mL | 0.2 cc | Centric Luer Slip |
Pack of 100 Each for $65.21
Case of 20 Pack for $1,304.20
|
|
|||
14-817-32
|
Air-Tite™
A20HJ |
20 mL | 1 cc | Eccentric Luer Tip |
Pack of 100 Each for $150.21
Case of 8 Pack for $1,201.68
|
|
|||
14-817-30
|
Air-Tite™
A10HJ |
10 mL | 0.5 cc | Eccentric Luer Tip |
Pack of 100 Each for $80.69
Case of 12 Pack for $968.28
|
|
|||
14-817-44
|
Air-Tite™
ABC10LS |
10 mL | 0.05 cc | Centric Luer Slip |
Pack of 100 Each for $81.97
Case of 19 Pack for $1,557.43
|
|
|||
14-817-51
|
Air-Tite™
ABC20LS |
20 mL | 0.05 cc | Centric Luer Slip |
Pack of 100 Each for $144.24
Case of 10 Pack for $1,442.40
|
|
|||
Description
These Air-Tite, all plastic syringes are available compatible with a variety of needle styles. (Needles are sold separately.) Ideal for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures.
- Needle tips not included and must be purchased separately
- Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
- Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
- Luer Tip, Luer-Slip, or catheter tip compatibility
- Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
- Positive safety stop prevents accidental spills
- FDA registered
- Dose saver design with low dead space plug on the piston minimizes waste.
- Catheter syringe has a large opening similar to a nozzle and is usable with catheters.
Specifications
FDA, CE, ISO 9001, DEHP Free | |
For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures | |
Henke-Ject™ Syringe |
Plastic | |
Yes | |
Cone |