Air-Tite™ All-Plastic Henke-Ject™ Syringes Edge
Edge
Inert syringes for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures. Air-Tite All Plastic Henke-Ject™ syringes are available compatible with a variety of needle styles. Needles are sold separately.
Supplier:  Air-Tite™ AL5HJ
 View more versions of this product
Catalog No. 14-817-29
Edge
Explore available promotions
Description
Description
These Air-Tite, all plastic syringes are available compatible with a variety of needle styles. (Needles are sold separately.) Ideal for use in gas or liquid chromatography, in vitro fertilization (IVF), embryo-transfer, and various laboratory procedures.
  • Needle tips not included and must be purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic, these syringes are ideal for chromatography, in vitro fertilization and embryo transfer
  • Luer Tip, Luer-Slip, or catheter tip compatibility
  • Polypropylene barrel and polyethylene plunger are chemically inert and noncytotoxic; needles contain no latex, synthetic rubber, silicone oil, or styrene
  • Positive safety stop prevents accidental spills
  • FDA registered
Order Info
Individually wrapped in convenient sterile strip packaging.
Specifications
Specifications
FDA, CE, ISO 9001, DEHP Free
For nuclear medicine, amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures
Henke-Ject™ Syringe
5 mL
Cone
Yes
Plastic
Yes
0.2 cc
Norm-Ject
Luer Tip
Sterile