missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reagent Alcohol, Fisher Chemical™

Specially Denatured Alcohol for histologic use.

Supplier:  Thermo Fisher Scientific BPA962P4

 View more versions of this product

Catalog No. A962P-4



Explore available promotions

Description

Description

Note:  Federal registration is required to purchase sizes greater than 4L.
Specifications

Specifications

Reagent Alcohol
64-17-5,67-63-0,67-56-1
78.5°C
12.8°C (55°F)
Liquid
Assay: Ethanol (88 to 91%), Methanol (4.0 to 5.0% v/v), Isopropyl Alcohol (4.5 to 5.5%)
C2H6O
44mmHg
0.10% Max.
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
Histological
-114.1°C
0.8g/cm³
4 L
Poly Bottle
Reagent
0.782 to 0.793
MFCD00003568
1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
Anhydrous Ethyl Alcohol 90% ±1% v/v; Methyl Alcohol approx. 5% v/v; 2-Propanol approx. 5% v/v

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Harmful by inhalation, in contact with skin and if swallowed. Vapor harmful. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions