Learn More
Reagent Alcohol, Fisher Chemical™
Specially Denatured Alcohol for histologic use.
$229.83 - $6882.58
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A962-4
|
Fisher Chemical
BPA9624 |
4 L | Glass Bottle |
Each for $342.58
Case of 4 Each for $1,370.32
|
|
||||
A962P-4
|
Fisher Chemical
BPA962P4 |
4 L | Poly Bottle |
Each for $229.83
Case of 4 Each for $919.32
|
|
||||
A962-200
|
Thermo Fisher Scientific
BPA962200 |
200 L | Steel Drum |
Each for $6,882.58
|
|
||||
Description
Note: Federal registration is required to purchase sizes greater than 4L.Chemical Identifiers
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
-114.1°C | |
0.8g/cm³ | |
4 L | |
Glass Bottle | |
Assay: Ethanol (88 to 91%), Methanol (4.0 to 5.0% v/v), Isopropyl Alcohol (4.5 to 5.5%) | |
C2H6O | |
44mmHg | |
0.10% Max. | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
Reagent | |
Reagent Alcohol |
64-17-5,67-63-0,67-56-1 | |
78.5°C | |
12.8°C (55°F) | |
Liquid | |
0.782 to 0.793 | |
MFCD00003568 | |
1170 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
Anhydrous Ethyl Alcohol 90% ±1% v/v; Methyl Alcohol approx. 5% v/v; 2-Propanol approx. 5% v/v |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Harmful by inhalation, in contact with skin and if swallowed. Vapor harmful. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid