Allegro™ Respirator Cleaning Pads
Generously sized pads extend wear between complete cleanings.
Supplier:  Allegro™ CAN1001
 View more versions of this product
Catalog No. 19-049883
Explore available promotions
Description
Description
  • Designed specifically for use on rubber respirator facepieces and other personal safety equipment
  • Available with 70% isopropanol or alcohol-free (with benzalkonium chloride) to clean even the most sensitive rubber facepiece
  • Individually foil packed in quantities of 50/Pk. or 100/Pk.
Specifications
Specifications
Respirator Wipes
Alcohol
100/Pk.
8 in.
5 in.
1001
Respirator facepiece
8 x 5 in. (13 x 20 cm)
13 cm
20 cm