missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allegro™ Respirator Cleaning Pads

Generously sized pads extend wear between complete cleanings.

$24.75 - $30.14

Specifications

Product Type Respirator Wipes
For Use With (Equipment) Respirator facepiece
Length (English) 8 in.
Length (Metric) 13 cm
View More Specs

Products 4
Catalog Number Mfr. No. Solution Type Quantity Price Quantity  
Catalog Number Mfr. No. Solution Type Quantity Price Quantity  
19-088-523
View Documents
Allegro™
100105
Alcohol 50/Pk.
Pack of 50 Each for $27.81
 
19-088-524
View Documents
Allegro™
300105
Alcohol-free 50/Pk.
Pack of 50 Each for $30.12
 
19-049883
View Documents
Allegro™
CAN1001
Alcohol 100/Pk.
Pack of 100 Each for $30.14
 
19-038-507
View Documents
Allegro™
CAN3001
Alcohol-free 100/Pk.
Pack of 100 Each for $24.75
 
Description

Description

  • Designed specifically for use on rubber respirator facepieces and other personal safety equipment
  • Available with 70% isopropanol or alcohol-free (with benzalkonium chloride) to clean even the most sensitive rubber facepiece
  • Individually foil packed in quantities of 50/Pk. or 100/Pk.

Specifications

Specifications

Respirator Wipes
Respirator facepiece
8 in.
13 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'