Allegro™ Respirator Cleaning Pads
Generously sized pads extend wear between complete cleanings.
Supplier: Allegro™ 100105
Description
- Designed specifically for use on rubber respirator facepieces and other personal safety equipment
- Available with 70% isopropanol or alcohol-free (with benzalkonium chloride) to clean even the most sensitive rubber facepiece
- Individually foil packed in quantities of 50/Pk. or 100/Pk.
Specifications
Respirator Wipes | |
Alcohol | |
50/Pk. | |
8 in. | |
11 in. |
1001-05 | |
Respirator facepiece | |
8 x 11 in. (13 x 28 cm) | |
13 cm | |
28 cm |