Synthware™ Allihn Condenser with Removable Hose Connections
Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint
Supplier:  Synthware™ C621200
 View more versions of this product
Catalog No. 31500798
Explore available promotions
Description
Description
Specifications
Specifications
24/40
Organic Synthesis
200 mm
Borosilicate Glass
0
Allihn Condenser
1 Ea.