missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl bromide, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 102900050

 View more versions of this product

Catalog No. AC102900050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Allyl bromide
106-95-6,75-56-9
100.0
Colorless
70.0°C to 71.0°C
Authentic
The liquid has a pungent odor
C3H5Br
H2C=CHCH2Br
5 L
13,219
15,279
Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide
C=CCBr
120.98
7841
99%
Stabilized
98.5
-119.0°C
1.3900g/mL
−1°C
98.5% min. (GC)
Metal Can
1.4680 to 1.4700
MFCD00000244
01,201
1.39
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
BHELZAPQIKSEDF-UHFFFAOYSA-N
3-bromoprop-1-ene
300ppm propylene oxide
120.98
Liquid

Chemical Identifiers

106-95-6
120.98
BHELZAPQIKSEDF-UHFFFAOYSA-N
7841
C=CCBr
C3H5Br
MFCD00000244
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
3-bromoprop-1-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life.
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 203-446-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only