Learn More
Allyl bromide, 99%, stabilized, Thermo Scientific Chemicals
$113.77 - $797.01
Chemical Identifiers
CAS | 106-95-6 |
---|---|
Molecular Formula | C3H5Br |
Molecular Weight (g/mol) | 120.98 |
MDL Number | MFCD00000244 |
InChI Key | BHELZAPQIKSEDF-UHFFFAOYSA-N |
Synonym | allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide |
PubChem CID | 7841 |
IUPAC Name | 3-bromoprop-1-ene |
SMILES | C=CCBr |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC102902500
|
Thermo Scientific Chemicals
102902500 |
250 mL | Glass bottle |
N/A
|
|
||||
AC102905000
|
Thermo Scientific Chemicals
102905000 |
500 mL | Glass bottle |
N/A
|
|
||||
AC102900050
|
Thermo Scientific Chemicals
102900050 |
5 L | Metal Can |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
106-95-6 | |
120.98 | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
7841 | |
C=CCBr |
C3H5Br | |
MFCD00000244 | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
3-bromoprop-1-ene |
Specifications
106-95-6,75-56-9 | |
100.0 | |
Colorless | |
70.0°C to 71.0°C | |
Authentic | |
The liquid has a pungent odor | |
C3H5Br | |
H2C=CHCH2Br | |
250 mL | |
13,219 | |
15,279 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide | |
C=CCBr | |
120.98 | |
7841 | |
99% | |
Allyl bromide, Stabilized |
98.5 | |
-119.0°C | |
1.3900g/mL | |
−1°C | |
98.5% min. (GC) | |
Glass bottle | |
1.4680 to 1.4700 | |
MFCD00000244 | |
01,201 | |
1.39 | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
3-bromoprop-1-ene | |
300ppm propylene oxide | |
120.98 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life.
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 203-446-6
RUO – Research Use Only