Learn More
Allyl chloroformate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221740500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
Allyl chloroformate | |
96.0 | |
Brown-Red to Yellow | |
109.0°C to 110.0°C | |
Authentic | |
Glass bottle | |
1.4213 to 1.4233 | |
MFCD00000648 | |
03, 12 | |
1.13 | |
Solubility in water: decomposes | |
C=CCOC(=O)Cl | |
120.54 | |
120.54 | |
Liquid |
2937-50-0 | |
100.0 | |
1.1300g/mL | |
31°C | |
96% min. (GC) | |
C4H5ClO2 | |
ClCO2CH2CH=CH2 | |
50 mL | |
12,15; 13,9 | |
allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
prop-2-enyl carbonochloridate | |
18052 | |
97% |
Chemical Identifiers
2937-50-0 | |
120.54 | |
CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
18052 | |
C=CCOC(=O)Cl |
C4H5ClO2 | |
MFCD00000648 | |
allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
prop-2-enyl carbonochloridate |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 220-916-6
RUO – Research Use Only