Learn More
Allyl chloroformate, 97%
CAS: 2937-50-0 | C4H5ClO2 | 120.54 g/mol
$240.11 - $463.37
Chemical Identifiers
| CAS | 2937-50-0 |
|---|---|
| Molecular Formula | C4H5ClO2 |
| Molecular Weight (g/mol) | 120.54 |
| MDL Number | MFCD00000648 |
| InChI Key | CAEWJEXPFKNBQL-UHFFFAOYSA-N |
| Synonym | allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech |
| PubChem CID | 18052 |
| IUPAC Name | prop-2-enyl carbonochloridate |
| SMILES | C=CCOC(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC221740500
|
Thermo Scientific Chemicals
221740500 |
50 mL | Glass bottle |
Each for $240.11
|
|
||||
|
AC221741000
|
Thermo Scientific Chemicals
221741000 |
100 mL | Glass bottle |
Each for $463.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
| 2937-50-0 | |
| 120.54 | |
| CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
| 18052 | |
| C=CCOC(=O)Cl |
| C4H5ClO2 | |
| MFCD00000648 | |
| allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
| prop-2-enyl carbonochloridate |
Specifications
| 2937-50-0 | |
| 100.0 | |
| 1.1300g/mL | |
| 31°C | |
| 96% min. (GC) | |
| C4H5ClO2 | |
| ClCO2CH2CH=CH2 | |
| 50 mL | |
| 12,15; 13,9 | |
| allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
| CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
| prop-2-enyl carbonochloridate | |
| 18052 | |
| 97% | |
| Allyl chloroformate |
| 96.0 | |
| Brown-Red to Yellow | |
| 109.0°C to 110.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4213 to 1.4233 | |
| MFCD00000648 | |
| 03, 12 | |
| 1.13 | |
| Solubility in water: decomposes | |
| C=CCOC(=O)Cl | |
| 120.54 | |
| 120.54 | |
| Liquid |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 220-916-6
RUO – Research Use Only