Learn More
Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 215870025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allyl methacrylate | |
0.05% max. | |
0.9300g/mL | |
37°C | |
97.5% min. (GC) | |
C7H10O2 | |
H2C=C(CH3)CO2CH2CH=CH2 | |
2.5 L | |
0.93 | |
Solubility in water: 4g/L (20°C). Other solubilities: miscible with almost all organic solvents | |
CC(=C)C(=O)OCC=C | |
126.16 | |
7274 | |
126.15 | |
Liquid |
96-05-9 | |
-65.0°C | |
144.0°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.4370 | |
MFCD00008592 | |
02,III,129 | |
allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech | |
FBCQUCJYYPMKRO-UHFFFAOYSA-N | |
prop-2-enyl 2-methylprop-2-enoate | |
200 to 250ppm MEHQ | |
1 mPa.s (20°C) | |
98% |
Chemical Identifiers
96-05-9 | |
126.16 | |
FBCQUCJYYPMKRO-UHFFFAOYSA-N | |
7274 | |
CC(=C)C(=O)OCC=C |
C7H10O2 | |
MFCD00008592 | |
allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech | |
prop-2-enyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure in contact with skin.
Flammable liquid and vapor.
Very toxic to aqu
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 202-473-
RUO – Research Use Only