missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allyl methacrylate, 98%, stabilized, Thermo Scientific Chemicals

$149.27 - $629.58

Chemical Identifiers

CAS 96-05-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00008592
InChI Key FBCQUCJYYPMKRO-UHFFFAOYSA-N
Synonym allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech
PubChem CID 7274
IUPAC Name prop-2-enyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215875000
SDS View Documents
Thermo Scientific Chemicals
215875000
500 mL Glass bottle
Each for $149.27
 
AC215870025
SDS View Documents
Thermo Scientific Chemicals
215870025
2.5 L Glass bottle
Each for $629.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-05-9
126.16
FBCQUCJYYPMKRO-UHFFFAOYSA-N
7274
CC(=C)C(=O)OCC=C
C7H10O2
MFCD00008592
allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech
prop-2-enyl 2-methylprop-2-enoate

Specifications

96-05-9
-65.0°C
144.0°C
Authentic
Glass bottle
1.4350 to 1.4370
MFCD00008592
02,III,129
allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech
FBCQUCJYYPMKRO-UHFFFAOYSA-N
prop-2-enyl 2-methylprop-2-enoate
200 to 250ppm MEHQ
1 mPa.s (20°C)
98%
Allyl methacrylate
0.05% max.
0.9300g/mL
37°C
97.5% min. (GC)
C7H10O2
H2C=C(CH3)CO2CH2CH=CH2
500 mL
0.93
Solubility in water: 4g/L (20°C). Other solubilities: miscible with almost all organic solvents
CC(=C)C(=O)OCC=C
126.16
7274
126.15
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure in contact with skin.
Flammable liquid and vapor.
Very toxic to aqu

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 202-473-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only