missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allylbenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102895000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allylbenzene | |
Colorless to Yellow | |
156.0°C to 157.0°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.5120 | |
500 mL | |
0.89 | |
Solubility in water: insoluble | |
C=CCC1=CC=CC=C1 | |
118.18 | |
118.18 | |
Liquid |
300-57-2 | |
0.8900g/mL | |
33°C | |
98% | |
C9H10 | |
C6H5CH2CH=CH2 | |
05,484 | |
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene | |
HJWLCRVIBGQPNF-UHFFFAOYSA-N | |
prop-2-enylbenzene | |
9309 | |
98% |
Chemical Identifiers
300-57-2 | |
118.18 | |
allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene | |
prop-2-enylbenzene |
C9H10 | |
HJWLCRVIBGQPNF-UHFFFAOYSA-N | |
9309 | |
C=CCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 206-095-7
RUO – Research Use Only